Sodium bisulfite, granular FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Sodium hydrogen sulphite

S1172-500GM S1172-2.5KG S1172-12KG S1172-45KGBL
95035-732EA 113.46 USD
95035-732 95035-730 700001-456 89079-000
Sodium bisulfite, granular FCC
Sodium bisulphite

Sodium Bisulfite, Granular, FCC is used as an antioxidant to preserve the aromas and flavors. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: HNaO₃S
MW: 104.06 g/mol
Melting Pt: 300 °C
Density: 2.476 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7631-90-5
EINECS: 231-548-0
UN: 2693
ADR: 8,III
Merck Index: 12,08731

Order Now

Specification Test Results

Assay 58.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR