Sodium aluminate, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Sodium Aluminium oxide, Aluminum sodium oxide, Aluminium sodium dioxide, Sodium aluminum oxide

S1120-500GM S1120-2.5KG
700001-980EA 133.75 USD
700001-980 700001-978
Sodium aluminate, Technical Grade
Sodium aluminate

Sodium Aluminate, Technical is an inorganic chemical that is used as in water softening systems, as a coagulant aid to improve flocculation, and for removing dissolved silica and phosphates. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Formula: NaAlO₂
MW: 81.97 g/mol
Melting Pt: 1800 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00801010
CAS Number: 1302-42-7
EINECS: 215-100-1
UN: 2812
ADR: 8,
Merck Index: 12,08715

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR