Sodium alginate NF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Alginic acid sodium salt, Algin
S1118-50KGBL
S1118-12KG
S1118-2.5KG
S1118-500GM
700013-080EA
3982.83
USD
700013-080
700013-078
95035-668
95035-670
Sodium alginate NF
Sodium alginate
Sodium Alginate, NF is a gum that has no discernible flavor and is used in the food and pharmaceutical industries as an emulsifier and to increase viscosity. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Storage Temperature:
Ambient |
MDL Number:
MFCD00081310 CAS Number: 9005-38-3 |
Specification Test Results
Assay | 90.8% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...