Disodium dihydrogen diphosphate ≥93% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Disodium pytophosphate, Sodium pyrophosphate, Disodium Dihydrogen Pyrophosphate, Sodium Acid Pyrophosphate

S1099-45KGBL S1099-500GM S1099-12KG S1099-2.5KG
700001-772EA 941.45 USD
700001-772 95035-662 700001-770 95035-660
Disodium dihydrogen diphosphate ≥93% FCC suitable for use as excipient
Disodium dihydrogen diphosphate

Sodium Acid Pyrophosphate, FCC is leavening agent found in baking powders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₂Na₂O₇P₂
Density: 2.311 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014246
CAS Number: 7758-16-9
EINECS: 231-835-0
Merck Index: 12,08713

Order Now

Specification Test Results

Assay 93% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR