Silver nitrate 99, crystals ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
Silver Nitrate, Crystal, Reagent, ACS is an inorganic compound with the formula AgNO₃. Silver nitrate is an extremely versatile analytical laboratory reagent for both the qualitative and quantitative determination of halides, carbonates, hydroxides, sulfides and phosphates. Spectrum provides solid Reagent ACS grade silver nitrate for preparation of analytical laboratory reagent solutions. As an ACS grade Reagent, Spectrum Chemical manufactured Silver Nitrate is used as the quality reagent in standard testing to which other substances are graded. It has met the toughest regulatory standards for quality and purity
Formula:
AgNO₃ MW: 169.87 g/mol Boiling Pt: 444 Melting Pt: 210 Density: 4.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1 II REACH: 01-2119513705-43 Merck Index: 12,08661 |
|
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...