Silver chloride ≥99%, granular, Reagent Grade for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
S1055-100GM
S1055-25GM
S1055-500GM
700002-070EA
1236
USD
700002-070
700002-074
700002-076
Silver chloride ≥99%, granular, Reagent Grade for synthesis
Silver chloride
Silver Chloride, Granular, Reagent is a chemical compound of silver and chlorine, with a molecular formula AgCl. There are many applications for SILVER CHLORIDE, including those in electrochemistry, infra-red technology, photochromic lenses, photographic paper, as well as bandages. SILVER CHLORIDE, GRANULAR, REAGENT is an analytical reagent used in some laboratory tests to determine the purity of other products and whether they can be graded pure enough for use in cosmetics, personal care, pharmaceutical or food and beverages
Formula:
AgCl MW: 143.32 g/mol Boiling Pt: 1554 Melting Pt: 455 Density: 5.560 Storage Temperature: Ambient |
MDL Number:
MFCD00003399 CAS Number: 7783-90-6 EINECS: 232-033-3 Merck Index: 13,08582 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...