L(+)-Serine ≥98.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

S1021-1KG S1021-25GM S1021-100GM
95034-068EA 1356.5 USD
95034-068 95034-070 95034-066
L(+)-Serine ≥98.5% FCC
L(+)-Serine

L-Serine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 215…225 °C
Density: 1.6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR