D-(-)-Ribose ≥97%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.

RI105-100GM RI105-25KGBL RI105-1KGBL RI105-25GM RI105-5GM
700009-708EA 266.35 USD
700009-708 700009-714 700009-710 700009-712 700009-716
D-(-)-Ribose ≥97%, Reagent Grade
D-(-)-Ribose

D-Ribose, Reagent is known to form part of the backbone of RNA and related to deoxyribose which is also found in DNA. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR