Rhodamine 6G

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

RH120-1KGBL RH120-100GM RH120-25GM RH120-5GM
700010-198EA 1214.3 USD
700010-198 700010-196 700010-200 700010-202
Rhodamine 6G
Rhodamine 6G

Rhodamine 6G, also known as Rhodamine 590, is a fluorescent rhodamine dye used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry, fluorescence correlation spectroscopy and ELISA.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR