Rhodamine B
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
RH115-100GM
RH115-25GM
RH115-500GM
700008-556EA
116
USD
700008-556
700008-558
700008-560
Rhodamine B
Rhodamine B
Rhodamine B, also known as Rhodamine 610, is used as a tracer dye within water to determine the direction and rate of transport and flow.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂₈H₃₁ClN₂O₃ MW: 479.02 g/mol Melting Pt: 199 - 201 °C Density: 1.31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011931 CAS Number: 81-88-9 EINECS: 201-383-9 Merck Index: 13,08266 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...