(-)-Quinine, anhydrous ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

QU108-1KGBL QU108-25GM
700009-572EA 3093.4 USD
700009-572 700009-574
(-)-Quinine, anhydrous ≥98%
(-)-Quinine

Quinine, Anhydrous is an antiarrhythmic agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₂₄N₂O₂
MW: 324.42 g/mol
Melting Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR