(±)-1-Phenoxy-2-propanol ≥95%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.

PR132-4LT PR132-25ML PR132-20LT
700003-554EA 799.1 USD
700003-554 700003-556 700003-552 700003-550
(±)-1-Phenoxy-2-propanol ≥95%, Reagent Grade
(±)-1-Phenoxy-2-propanol

Propylene Phenoxytol, Reagent

Formula: C₆H₅OCH₂CH(OH)CH₃
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 11.4 °C
Density: 1.0665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016861
CAS Number: 770-35-4
EINECS: 212-222-7

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR