Polysorbate 85
Supplier: SPECTRUM CHEMICAL MFG CORP
PO140-20LT
PO140-4LT
700009-832EA
225.16
USD
700009-832
700009-828
700009-830
Polysorbate 85
Polysorbate 85
Polysorbate 85, also known as sorbitan monooleate, is a nonionic emulsifier and surfactant and used in cosmetics and foods.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
1.028 Storage Temperature: Ambient |
MDL Number:
MFCD01779641 CAS Number: 9005-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...