Polysorbate 40 NF

Supplier: SPECTRUM CHEMICAL MFG CORP

PO134-20LT PO134-4LTGL PO134-500ML
700011-054EA 1889.15 USD
700011-054 95035-044 95035-046
Polysorbate 40 NF
Polysorbate 40

Polysorbate 40, NF is used to solubilize essential oils. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₆₂H₁₂₂O₂₆
Density: 1.083
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR