Polyethylene glycol 3,350 USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: PEG 3,350, PEG 3350

PO125-12KG PO125-45KGBL PO125-500GM PO125-2.5KG
76181-244EA 1158.82 USD
76181-244 76181-246 76181-300 76181-302
Polyethylene glycol 3,350 USP
Polyethylene glycol 3,350

Polyethylene Glycol 3350, USP acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR