Polyethylene glycol 1,500 NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: PEG 1500, PEG 1,500

PO120-500GM PO120-12KG PO120-2.5KG PO120-45KGBL
95034-990EA 124 USD
95034-990 700012-606 95034-988 89078-936
Polyethylene glycol 1,500 NF
Polyethylene glycol 1,500

Polyethylene Glycol 1500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 44…48 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR