Polyethylene glycol 600 NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethylene glycol polymer average Mn 600, PEG 600

PO115-20LT PO115-200LTB PO115-2.5LTG PO115-500MLG PO115-2.5LTG
700012-600EA 1165.85 USD
700012-600 700012-598 95035-010 95035-012 700012-596
Polyethylene glycol 600 NF
Polyethylene glycol 600

Polyethylene Glycol 600, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: H(OCH₂CH₂)nOH
MW: 600 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 17…22 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR