Phenyl salicylate ≥98%, crystals for synthesis, purified
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Phenyl 2-hydroxybenzoate, Phenyl Salicylate
PH160-125GM
PH160-500GM
PH160-12KG
PH160-2.5KG
PH160-50KGBL
700006-430EA
177.05
USD
700006-430
700006-436
700006-432
700006-434
700006-438
Phenyl salicylate ≥98%, crystals for synthesis, purified
Phenyl salicylate
Phenyl Salicylate, Crystal, Purified is used in the manufacture of some polymers, lacquers, adhesives, waxes and polishes.
The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol Boiling Pt: 307 Melting Pt: 41…43 °C Density: 1.307 g/cm³ (20 °C) Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002213 CAS Number: 118-55-8 EINECS: 204-259-2 UN: 3077 ADR: 9 III Merck Index: 12,07464 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...