Patent Blue VF (Disulphine blue) for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: acid Blue1, Patent Blue, Patent Blue VF, Hydrogen [4-[4-(diethylamino)-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium, sodium salt, Patent blue VF sodium salt

PA135-100GM PA135-25GM PA135-500GM
700010-130EA 197.53 USD
700010-130 700010-132 700010-134
Patent Blue VF (Disulphine blue) for synthesis
Patent Blue VF (Disulfine blue)

Patent Blue, also known as Food Blue 5, is a blue synthetic triphenylmethane dye which is used as food coloring.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₇H₃₁N₂NaO₆S₂
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012119
CAS Number: 129-17-9
EINECS: 204-934-1
Merck Index: 12,09161

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR