1,3-Propanediol ≥98%

Supplier: Spectrum Chemical Mfg. Corp.

P2139-100ML P2139-500MLG
700003-006EA 90.36 USD
700003-006 700003-008
1,3-Propanediol ≥98%
1,3-Propanediol

1,3-Propanediol, an organic compound, has a slightly sweet taste and chemically classed as a diol. It is miscible with several solvents including chloroform, water and acetone. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: HO(CH₂)₃OH
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: -32 °C
Density: 1.0597 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002949
CAS Number: 504-63-2
EINECS: 207-997-3
Merck Index: 12,09845

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR