2-Phenylethyl acetate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate
P2013-200LTB
P2013-1LT
P2013-4LT
P2013-250ML
700004-622EA
6844.8
USD
700004-622
700004-620
700004-626
700004-624
2-Phenylethyl acetate
2-Phenylethyl acetate
Phenylethyl Acetate is used as fragrance ingredient for home cleaning products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 233 °C (1013 hPa) Melting Pt: –31 °C Density: 1.0286 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008720 CAS Number: 103-45-7 EINECS: 203-113-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...