L(+)-Potassium hydrogen tartrate ≥99% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

P1801-12KG P1801-2.5KG P1801-500GM P1801-50KGBL
700006-136EA 1522.65 USD
700006-136 95035-104 95035-106 89078-950
L(+)-Potassium hydrogen tartrate ≥99% USP
L(+)-Potassium hydrogen tartrate

Potassium Bitartrate, USP is used a stabilizer and pH control agent. 


All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: KOOC(CHOH)₂COOH
MW: 188.18 g/mol
Melting Pt: 250 °C
Density: 1.943 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR