Potassium sulfate ≥99% FCC

Supplier: Spectrum Chemical Mfg. Corp.

P1589-12KG P1589-50KGBL P1589-500GM P1589-2.5KG P1589-125GM
700009-908EA 1344.05 USD
700009-908 89078-976 95035-238 95035-236 95035-234
Potassium sulfate ≥99% FCC
Potassium sulphate

Potassium Sulfate, FCC is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: K₂SO₄
Boiling Pt: 1698 °C (1013 hPa)
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR