L-Phenylalanine ≥98.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
P1582-5KGBL
P1582-100GM
P1582-1KG
P1582-25GM
700007-408EA
11512
USD
700007-408
95034-048
95034-050
95034-052
L-Phenylalanine ≥98.5% USP
L-Phenylalanine
Phenylalanine, USP is an amino acid, a building block of protein.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Boiling Pt: 313 °C (1013 hPa) Melting Pt: 173 °C Density: 0.754 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...