Pyrogallol, crystals ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

P1460-12KGBL P1460-50KGBL P1460-25GM P1460-500GM P1460-125GM
700003-316EA 7801.3 USD
700003-316 700003-322 700003-318 700003-320 700003-314
Pyrogallol, crystals ACS
Pyrogallol

Pyrogallol, Crystal, Reagent, ACS, also known as Pyrogallic acid, is one of three isomeric benzenetriols that is used for oxygen absorption in gas analysis as well as in hair dying.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1.453 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,08090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR