(±)-1-Methoxy-2-propanol ≥97%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol

P1438-20LT P1438-1LT P1438-4LTGL P1438-55GLBL
700003-530EA 1210.45 USD
700003-530 700003-528 700003-532 700003-534
(±)-1-Methoxy-2-propanol ≥97%
(±)-1-Methoxy-2-propanol

Propylene Glycol Methyl Ether is an organic compound that has no color, no odor, is a transparent viscous liquid and has a lightly sweet tatse. It is miscible and hygroscopic with water, acetone and chloroform. Nearly half of all propylene glycol made is used for the fabrication of unsaturated polyester resin. It is also used in food and tabacco products as a humectant, preservative and solvent.

Formula: CH₃CH(OH)CH₂OCH₃
MW: 90.12 g/mol
Boiling Pt: 119.6 °C (1013 hPa)
Melting Pt: –97 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR