L(-)-Proline USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH

P1434-25GM P1434-25KGBL P1434-100GM P1434-1KG P1434-5KG
95034-064EA 136.59 USD
95034-064 89078-846 95034-060 95034-062 700007-406
L(-)-Proline USP
L(-)-Proline

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220 - 222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR