1-Propanol ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
P1430-4LTGL
P1430-500MLG
700002-642EA
333.07
USD
700002-642
700002-644
1-Propanol ACS
1-Propanol
n-Propyl Alcohol, Reagent, ACS is a colorless liquid that is also known as propanol and is a primary alcohol. It is used in the laboratory primarily as an analytical solvent. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness
Formula:
H₃CCH₂CH₂OH Boiling Pt: 97 °C (1013 hPa) Melting Pt: -127 °C Density: 0.8053 g/cm³ (20 °C) Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3,II Merck Index: 13,07934 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...