Dipotassium hydrogen orthophosphate trihydrate ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
P1385-12KG
P1385-125GM
P1385-2.5KG
P1385-500GM
700001-700EA
1963.6
USD
700001-700
700001-698
700001-702
700001-706
Dipotassium hydrogen orthophosphate trihydrate ≥98%, Reagent Grade
Dipotassium hydrogen orthophosphate trihydrate
Potassium Phosphate Dibasic, Trihydrate, Reagent is a soluble salt that is commonly used as a buffering agent. As a Reagent Grade, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
K₂HPO₄·3H₂O MW: 228.22 g/mol Boiling Pt: ? 340 °C (1013 hPa) Density: 2.504 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149926 CAS Number: 16788-57-1 EINECS: 231-834-5 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...