Tripotassium orthophosphate ≥97% FCC suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Potassium phosphate tribasic, Potassium phosphate
Potassium Phosphate Tribasic, FCC is used as a food additive for its properties as an emulsifier, foaming agent, and whipping agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
K₃O₄P MW: 212.27 g/mol Melting Pt: 1340 Density: 2.564 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036295 CAS Number: 7778-53-2 EINECS: 231-907-1 Merck Index: 12,07830 |
Specification Test Results
Assay | 97% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...