di-Potassium oxalate monohydrate ≥98.5%, crystals ACS

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate

P1355-125GM P1355-12KG P1355-2.5KG P1355-500GM P1355-50KGBL
700005-770EA 126.15 USD
700005-770 700005-772 700005-774 700005-776 700005-778
di-Potassium oxalate monohydrate ≥98.5%, crystals ACS
di-Potassium oxalate monohydrate

Form: Crystal

Formula: (COOK)₂·H₂O
MW: 184.23 g/mol
Melting Pt: 356 °C
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,07820

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR