Potassium disulfite, crystals NF

Supplier: Spectrum Chemical Mfg. Corp.
Danger

P1346-2.5KG P1346-50KGBL P1346-500GM
700002-022EA 1150.59 USD
700002-022 95035-192 700002-024 95035-194
Potassium disulfite, crystals NF
Potassium disulphite

Kosher certified


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: K₂O₅S₂
Melting Pt: 190 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167605
CAS Number: 16731-55-8
EINECS: 240-795-3
Merck Index: 13,07729

Order Now

Specification Test Results

Assay 51.8% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR