Potassium fluoride dihydrate ≥99%, crystals, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
P1310-12KG
P1310-2.5KG
P1310-45KGBL
P1310-500GM
P1310-125GM
P1310-25GM
700001-008EA
5169.05
USD
700001-008
700001-010
700001-014
700001-016
700001-006
700001-012
Potassium fluoride dihydrate ≥99%, crystals, Reagent Grade
Potassium fluoride dihydrate
Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
FK·2H₂O MW: 94.13 g/mol Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 846 °C Density: 2.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149929 CAS Number: 13455-21-5 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...