Potassium fluoride, anhydrous ≥99% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
P1306-1KG
P1306-2.5KG
P1306-500GM
P1306-25GM
P1306-50KGBL
P1306-125GM
P1306-12KG
700000-996EA
493.83
USD
700000-996
700000-998
700001-002
700001-000
700001-004
700000-992
700000-994
Potassium fluoride, anhydrous ≥99% ACS
Potassium fluoride
Potassium Fluoride, Anhydrous, Reagent, ACS is a source of the fluoride ion, and is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 860 °C Density: 2.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 13,07714 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...