n-Pentane ≥98%, Reagent Grade
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Pentane, Reagent is a straight chain hydrocarbon with five carbon atoms. It is a colorless, nonflammable liquid that has a strong hydrocarbon odor. It is a component in fuels and it is also used as a solvent in laboratories. It has very similar properties to butanes and hexanes. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
H₃C(CH₂)₃CH₃ Boiling Pt: 36.1 °C (1013 hPa) Melting Pt: -130 °C Density: 0.626 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II Merck Index: 12,07255 |
|
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...