Orange G for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: CI 16230

OR105-100GM OR105-1KGBL OR105-25GM
700010-124EA 277.39 USD
700010-124 700010-126 700010-128
Orange G for synthesis
Orange G

Orange G is an azo dye used primarily as a histological stain.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141
Density: 0.45…0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR