Octyltrimethoxysilane
Supplier: Spectrum Chemical Mfg. Corp.
O1424-100GM
O1424-25GM
700008-236EA
232.05
USD
700008-236
700008-238
Octyltrimethoxysilane
Octyltrimethoxysilane
Formula:
C₁₁H₂₆O₃Si Density: 0,907 Storage Temperature: Ambient |
MDL Number:
MFCD00043053 CAS Number: 3069-40-7 EINECS: 221-338-7 |
Specification Test Results
Assay | 96% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...