Caprylic acid ≥97%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

O1003-4LTGL O1003-1LT O1003-200LTB O1003-20LTBL O1003-100ML
700005-302EA 635.8 USD
700005-302 700005-296 700005-298 700005-300 700005-294
Caprylic acid ≥97%, Reagent Grade
Caprylic acid

Octanoic Acid, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR