1-Octanesulfonic acid sodium salt monohydrate ≥99% for HPLC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate
1-Octanesulfonic Acid Sodium Salt, Monohydrate, Reagent HPLC is an ion pairing reagent which will enhance the retention of polar, ionizable compounds such as peptides and proteins. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity
The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₈H₁₇NaO₃S·1H₂O MW: 234.29 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007544 CAS Number: 207596-29-0 EINECS: 226-195-4 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...