L-Norvaline ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

N3049-1GM N3049-25GM N3049-5GM
700007-382EA 82.75 USD
700007-382 700007-384 700007-388
L-Norvaline ≥99%
L-Norvaline

L-(+)-Norvaline, also known as 2-Aminopentyric acid or alpha-Aminopentanoic acid, is an amino acid and a component of Bacillus subtilis, an antifungal peptide; it is also known to promote muscle growth and tissue regeneration.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR