3-Nitrophenol ≥99% for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: m-Nitrophenol

N2033-100GM N2033-25GM N2033-1KGBL
700003-358EA 984 USD
700003-358 700003-362 700003-360
3-Nitrophenol ≥99% for synthesis
3-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 271
Melting Pt: 94…95 °C
Density: 1.485 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007240
CAS Number: 554-84-7
EINECS: 209-073-5
UN: 1663
ADR: 6.1 III
Merck Index: 13,06652

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR