4-Nitrophenol ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
N1099-100GM
N1099-2.5KG
N1099-500GM
700003-376EA
153.55
USD
700003-376
700003-378
700003-380
4-Nitrophenol ≥98%, Reagent Grade
4-Nitrophenol
p-Nitrophenol, Reagent, also known as 4-nitrophenol, is used in the synthesis of paracetamol. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 279 °C (1013 hPa) Melting Pt: 110…114 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007331 CAS Number: 100-02-7 EINECS: 202-811-7 UN: 1663 ADR: 6.1,III Merck Index: 13,06654 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...