2-Nitrophenol ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: o-Nitrophenol

N1098-250KGB N1098-100GM N1098-25GM
700003-372EA 9091.8 USD
700003-372 700003-370 700003-374
2-Nitrophenol ≥99%
2-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 214.6 °C (1013 hPa)
Melting Pt: 43 - 45 °C
Density: 1.599 g/cm³ (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1,III
Merck Index: 12,06717

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR