Myristic acid ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

MY110-2.5KG MY110-45KGBL MY110-100GM MY110-12KG MY110-500GM
700005-282EA 610.55 USD
700005-282 700005-284 700005-278 700005-280 700005-286
Myristic acid ≥98%, Reagent Grade
Myristic acid

Myristic Acid, Reagent, also known as tetradecanoic acid, acts as a lipid anchor in biomembranes. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₄H₂₈O₂
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 51…54 °C
Density: 0.8622 g/cm³ (20 °C)
Flash Pt: >165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR