Mineral oil, white (petroleum) light NF

Supplier: Spectrum Chemical Mfg. Corp.
Warning

MI501-200LTB MI501-20LTPL MI501-4CS4 MI501-500MLG MI501-500MLGL MI501-4LTGL MI501-1LT
700000-468EA 2365.95 USD
700000-468 700000-470 95034-648 95034-652 700000-472 95034-650 95034-646
Mineral oil, white (petroleum) light NF
Mineral oil, white (petroleum)

Light Mineral Oil, NF is used as a vehicle and humectant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

SPECIFIC GRAVITY @ 25°C 0.818-0.880
VISCOSITY @40.0 °C 3.0-34.4 mm²/s
ACIDITY TO PASS TEST
READILY CARBONIZABLE SUBSTANCES TO PASS TEST
POLYCYCLIC AROMATIC HYDROCARBONS TO PASS TEST
SOLID PARAFFIN TO PASS TEST
SULFUR COMPOUNDS TO PASS TEST
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR