Methyl cellulose, 4000 mPa.S (2% H₂O) USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Cellulose methyl ether

ME137-2.5KG ME137-12KG ME137-500GM ME137-45KGBL
95034-478EA 1275.75 USD
95034-478 700013-046 95034-476 95034-480 700013-048
Methyl cellulose, 4000 mPa.S (2% H₂O) USP
Methyl cellulose

Methylcellulose, 4,000 cP, USP is a tablet and capsule disintegrant, a tablet binder, and as a viscosity-increasing excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Assay 26% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR