Methyl cellulose, 400 cP (C=2%, H₂O) USP
Supplier: Spectrum Chemical Mfg. Corp.
Methylcellulose, 400 cP, USP is a tablet and capsule disintegrant, a tablet binder, and as a viscosity-increasing excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
Specification Test Results
Assay | 26% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...