D-(+)-Mannose ≥99%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
MA170-500GM
MA170-100GM
MA170-25GM
700009-690EA
1050.45
USD
700009-690
700009-686
700009-688
D-(+)-Mannose ≥99%, Reagent Grade
D-(+)-Mannose
D-Mannose, Reagent is a crucial element in the glycosylation of certain proteins in human metabolism. It is a sugar monomer of the series of aldohexose of carbohydrates. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 376 °C (1013 hPa) Melting Pt: 133 °C Density: 1.539 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064122 CAS Number: 3458-28-4 EINECS: 222-392-4 Merck Index: 13,05772 |
Specification Test Results
Assay | 99% Min. |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...