(±)-Mandelic acid ≥98%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

MA160-100GM MA160-12KGBL MA160-2.5KG MA160-500GM
700006-354EA 84.1 USD
700006-354 700006-356 700006-358 700006-360
(±)-Mandelic acid ≥98%, Reagent Grade
(±)-Mandelic acid

DL-Mandelic Acid, Reagent is a white crystalline water soluble solid that is also soluble in polar organic solvents. Its an aromatic alpha hydroxy acid that is used a precursor to various drugs.


Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 117 - 120 °C
Density: 1.361 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064250
CAS Number: 90-64-2
EINECS: 210-277-1

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR