Magnesium hydrogen orthophosphate trihydrate ≥96% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
MA127-2.5KG
MA127-12KG
MA127-500GM
700001-738EA
491.8
USD
700001-738
700001-736
700001-740
Magnesium hydrogen orthophosphate trihydrate ≥96% FCC
Magnesium hydrogen orthophosphate trihydrate
Magnesium Phosphate, Dibasic, Trihydrate, FCC is listed on the FDA's Generally recognized as safe, or GRAS, list of substances. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
HMgO₄P·3H₂O MW: 174.33 g/mol Density: 2.131 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150390 CAS Number: 7782-75-4 EINECS: 231-823-5 |
Specification Test Results
Assay | 96% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...